Product Name
4-Amino-3,5-dichloro-2,6-difluoropyridine 99%
Product Number
324388
Product Brand
aldrich
CAS Number
2840-00-8
Molecular Weight
198.99
Test | Specification |
---|---|
APPEARANCE | WHITE CRYSTALS OR CRYSTALLINE POWDER |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE AND STANDARD AS |
ILLUSTRATED ON PAGE 1540D OF EDITION I, | |
VOLUME 3 OF "THE ALDRICH LIBRARY OF FT-IR | |
SPECTRA". | |
ELEMENTAL ANALYSIS | CARBON 29.7% - 30.6% |
NITROGEN 13.8% - 14.4% | |
HIGH PRESSURE LIQUID | 98.5% (MINIMUM) |
SOLUBILITY | 5% IN ETHANOL; CLEAR, COLORLESS SOLUTION |
APPROVED APRIL 5, 1999 JSB |