Product Name
Clethodim sulfone analytical standard
Product Number
92602
Product Brand
SIAL
CAS Number
111031-17-5
Molecular Weight
391.91
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Faint Yellow and White to Faint Yellow and Faint Beige |
APPEARANCE (FORM) | Viscous Liquid or Solid |
PURITY (HPLC AREA %) | ≥ 95.0 % |
IDENTIFICATION (LC-MS) | CORRESPONDS TO REQUIREMENTS |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |