Product Name
Martius Yellow sodium salt monohydrate 98%
Product Number
287814
Product Brand
ALDRICH
CAS Number
304655-86-5
Molecular Weight
274.16
Test | Specification |
---|---|
APPEARANCE | ORANGE POWDER |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
UV-VISIBLE SPECTRUM | C = 0.01G/L IN METHANOL |
E(430 +/- 3) NM = 16,000 (MINIMUM) | |
E(388 +/- 3) NM = 15,000 (MINIMUM) | |
E(257 +/- 3) NM = 11,000 (MINIMUM) | |
ELEMENTAL ANALYSIS | CARBON 42.8% - 44.9% |
NITROGEN 9.9% - 10.5% | |
THIN-LAYER | CONSISTENT WITH 98% PURITY. |
ICP ASSAY | 8.2% - 8.6% SODIUM |
SOLUBILITY | 0.1% IN METHANOL; CLEAR, VERY DARK YELLOW TO |
GOLD OR ORANGE SOLUTION | |
APPROVED OCTOBER 3, 2001 JSB |