Skip to Content
Merck
All Photos(1)

Key Documents

SY3H3D68231F

Synthonix Corporation

1H-1,2,4-Triazole-1-carboximidamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
£21.56
10 G
£26.94
25 G
£32.33

Vendor SKU: T39916-5G

£21.56


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
5 G
£21.56
10 G
£26.94
25 G
£32.33

About This Item

CAS Number:
Molecular Weight:
147.57
MDL number:

Vendor SKU: T39916-5G

£21.56


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

1H-1,2,4-triazole-1-carboximidamide hydrochloride

Inchi Code

1S/C3H5N5.ClH/c4-3(5)8-2-6-1-7-8;/h1-2H,(H3,4,5);1H

InChI key

JDDXNENZFOOLTP-UHFFFAOYSA-N

storage temp.

Room Temperature

Purity

97%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Crystal - Powder / Colour: White - Almost white

Product Link

https://synthonix.com/39916.html

Pictograms

Exclamation mark

Signal Word

Warning

Precautionary statements

MSDS

https://synthonix.com/39916.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service