Skip to Content
Merck
All Photos(1)

Key Documents

COLO 839

93051119, human blood, Lymphoblastoid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

COLO 839, 93051119

biological source

human blood

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian B lymphocyte

Cell Line Description

Derived from the peripheral blood of a 43 year old Caucasian male with widespread melanoma. The cells were transformed with defective EBV (B-95-W8).
COLO 839 (Sigma Catalogue number 93051119), COLO 858 (Sigma Catalogue number 93052613) and COLO 857 (Sigma Catalogue number 93051120) were all derived from the same 43 year-old male patient with malignant melanoma (see references below). This has been confirmed by STR profiling.

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,13
D13S317: 11,12
D16S539: 11,13
D5S818: 11,12
D7S820: 8,10
THO1: 9.3,10
TPOX: 8,11
vWA: 17,19

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 2-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service