ARLR3012A
PolarSafe™ Cardboard Freezer Box
with 196 place divider for PCR tubes
All Photos(3)
About This Item
UNSPSC Code:
41103000
NACRES:
NB.22
Recommended Products
manufacturer/tradename
Argos R3012A
technique(s)
PCR: suitable
General description
PolarSafe cryo/freezer boxes and dividers are made of sturdy white cardboard, with a moisture-repellent coating to protect form the effects of liquid nitrogen or condensation from refrigerated equipment. Boxes are sized to fit all standard stainless-steel and polycarbonate storage systems. Small holes on the bottom allow for safe drainage of liquid nitrogen or water. For use in mechanical freezers and liquid nitrogen tanks at temperature to -196°C.
Choice of boxes and dividers to accommodate tubes from 0.5 – 50mL.
Choice of boxes and dividers to accommodate tubes from 0.5 – 50mL.
Features and Benefits
Boxes are designed to hold single PCR tubes or strips of 8. The 1" height prevents the tubes from falling down inside the grids. Available for 64 or 196 tubes.
To maximize storage utility two of the boxes can be stacked inside each slot in the applicable stainless steel freezer racks.
To maximize storage utility two of the boxes can be stacked inside each slot in the applicable stainless steel freezer racks.
Legal Information
PolarSafe is a trademark of Argos Technologies, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)