17925
9-Carbazoleacetic acid
≥99.0% (T)
Synonym(s):
(9-Carbazolyl)acetic acid, 9-(Carboxymethyl)carbazole
About This Item
Assay
≥99.0% (T)
mp
191-194 °C
SMILES string
OC(=O)Cn1c2ccccc2c3ccccc13
InChI
1S/C14H11NO2/c16-14(17)9-15-12-7-3-1-5-10(12)11-6-2-4-8-13(11)15/h1-8H,9H2,(H,16,17)
InChI key
PBDMLLFEZXXJNE-UHFFFAOYSA-N
Gene Information
human ... FABP4(2167)
Application
Other Notes
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service