Skip to Content
Merck
All Photos(1)

Key Documents

08897

Sigma-Aldrich

D-Erythritol 4-phosphate lithium salt

≥95.0% (TLC)

Synonym(s):

(2R,3S)-2,3,4-Trihydroxybutylphosphate lithium salt, L-Erythritol 1-phosphate lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H11O7P · xLi+
CAS Number:
Molecular Weight:
202.10 (free acid basis)
UNSPSC Code:
12352201
NACRES:
NA.25

Assay

≥95.0% (TLC)

form

solid

color

white to off-white

storage temp.

−20°C

InChI

1S/C4H11O7P/c5-1-3(6)4(7)2-11-12(8,9)10/h3-7H,1-2H2,(H2,8,9,10)/t3-,4+/m0/s1

InChI key

QRDCEYBRRFPBMZ-IUYQGCFVSA-N

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Veiga-da-Cunha et al.
Journal of bacteriology, 175(13), 3941-3948 (1993-07-01)
It was recently observed that Leuconostoc oenos GM, a wine lactic acid bacterium, produced erythritol anaerobically from glucose but not from fructose or ribose and that this production was almost absent in the presence of O2. In this study, the
The enantiomorphic erythritol-4-phosphates.
MacDonald, D.L., et al.
Journal of the American Chemical Society, 78, 3720-3722 (1956)
Erythritol metabolism by Propionibacterium pentosaceum. The role of L-erythrulose 1-phosphate.
E J Wawszkiewicz
Biochemistry, 7(2), 683-687 (1968-02-01)
Distribution and structural form of phosphate ester groups in commercial starch phosphates.
Gramera, R.E., et al.
Cereal Chem., 43, 104-111 (1966)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service