Skip to Content
Merck
All Photos(2)

Key Documents

34373

Supelco

Siduron

PESTANAL®, analytical standard

Synonym(s):

1-(2-Methylcyclohexyl)-3-phenylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20N2O
CAS Number:
Molecular Weight:
232.32
Beilstein:
2725940
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC1CCCCC1NC(=O)Nc2ccccc2

InChI

1S/C14H20N2O/c1-11-7-5-6-10-13(11)16-14(17)15-12-8-3-2-4-9-12/h2-4,8-9,11,13H,5-7,10H2,1H3,(H2,15,16,17)

InChI key

JXVIIQLNUPXOII-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Siduron is a pre-emergence and post emergence phenylurea herbicide used for the control of annual grass weeds. Its mode of action involves the inhibition of photosynthesis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Siduron may be used as an analytical reference standard for the determination of the analyte in water, fruit juice, and crops by various chromatography techniques.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rong Jiang et al.
Chemosphere, 204, 483-491 (2018-04-22)
Siduron is a widely used herbicide in urban lawn and has been frequently detected in urban and suburban surface water. However, characteristics of its environmental behavior in soil are seldom reported. The combined pollution of heavy metals, especially for Cu
J W Newman et al.
Environmental health perspectives, 109(1), 61-66 (2001-02-15)
Substituted ureas and carbamates are mechanistic inhibitors of the soluble epoxide hydrolase (sEH). We screened a set of chemicals containing these functionalities in larval fathead minnow (Pimphales promelas) and embryo/larval golden medaka (Oryzias latipes) models to evaluate the utility of
Determination and quantitation of sulfonylurea and urea herbicides in water samples using liquid chromatography with electrospray ionization mass spectrometric detection.
Ayano E, et al.
Analytica Chimica Acta, 507(2), 211-218 (2004)
Determination of 266 pesticide residues in apple juice by matrix solid-phase dispersion and gas chromatography-mass selective detection
Chu XG, et al.
Journal of Chromatography A, 1063(1-2), 201-210 (2005)
Simultaneous determination of 15 phenylurea herbicides in rice and corn using HPLC with fluorescence detection combined with UV decomposition and post-column derivatization.
Mou R-X, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 875(2), 437-443 (2008)

Protocols

-methylcarbamate 10 μg/mL; Diuron; Propham; Siduron; Methiocarb, analytical standard; Linuron 10 μg/mL; Swep 10 μg/mL; Chlorpropham 10 μg/mL; Barban; Neburon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service