70590-M
BacVector Insect Cell Medium - Novagen
Serum-free medium optimized for growth of Sf9 Insect Cells
Synonym(s):
Serum-free Insect Cell Medium
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
manufacturer/tradename
Novagen®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
protect from light
shipped in
ambient
wet ice
storage temp.
2-8°C
General description
BacVector Insect Cell Medium is optimized for serum-free growth of Sf9 Insect Cells. This cell/medium combination is recommended for cotransfection of BacVector Triple Cut Virus DNA or BacMagic DNA and transfer plasmids to construct recombinant baculoviruses, and is compatible with Insect GeneJuice® Transfection Reagent. This medium is also recommended for transient expression from pIEx and pBiEx vector recombinants as well as plaque assays using the agarose overlay technique. Store in the dark at 4°C.
Warning
Toxicity: Irritant
Legal Information
GENEJUICE is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service