Skip to Content
Merck
All Photos(1)

Key Documents

64-201BP

Millipore

Visualizer Western Blot Detection Kit, mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
NB.23

species reactivity

all

technique(s)

western blot: suitable

detection method

chemiluminescent

Application

For use on immunoblots.

Packaging

Kit capacity: 50 mini-gel-sized blots for 2500cm2 of membrane

Components

Goat Anti-Mouse IgG, HRP conjugate Visualizer Detection Reagent B Visualizer Detection Reagent A

Quality

The reagents in this kit have been matched to optimize the range and sensitivity of detection using various antibodies in Western blot assays

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service