238597-M
Sodium sulfate
ACS reagent, ≥99.0%, anhydrous, powder
About This Item
Recommended Products
grade
ACS reagent
anhydrous
Assay
≥99.0%
form
powder
impurities
≤0.01% insolubles
≤5 ppm N compounds
loss
≤0.5% loss on ignition
pH
5.2-9.2
mp
884 °C (lit.)
density
2.68 g/mL at 25 °C (lit.)
anion traces
chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%
cation traces
Ca: ≤0.01%
Fe: ≤0.001%
K: ≤0.01%
Mg: ≤0.005%
heavy metals: ≤5 ppm (by ICP-OES)
SMILES string
[Na+].[Na+].[O-]S([O-])(=O)=O
InChI
1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2
InChI key
PMZURENOXWZQFD-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Other Notes
This product will be phased out towards early 2019. There is a replacement product available : 238597
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service