Skip to Content
Merck
All Photos(1)

Key Documents

1.51257

Supelco

Split collets for for MANU-CART HPLC cartridge holder

pkg of 2 pieces

Sign Into View Organizational & Contract Pricing

Select a Size

4 UNITS
£105.00

£105.00


Please contact Customer Service for Availability


Select a Size

Change View
4 UNITS
£105.00

About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

£105.00


Please contact Customer Service for Availability

Product Name

Split collets for manu-CART cartridge holder, pkg of 2 pieces

packaging

pkg of 2 pieces

Quality Level

technique(s)

HPLC: suitable

storage temp.

no temp limit

General description

(2 pieces)
The package contains: 4 pieces.

Analysis Note

visual control (for delivery): passes test

Legal Information

MANU-CART is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service