MAT333116449
Matrix Scientific
2,4-Diamino-N-phenylbenzamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2,4-diamino-N-phenylbenzamide
Inchi Code
1S/C13H13N3O/c14-9-6-7-11(12(15)8-9)13(17)16-10-4-2-1-3-5-10/h1-8H,14-15H2,(H,16,17)
InChI key
DLHWNBZHYWNPIG-UHFFFAOYSA-N
Country of Origin
US
Product Link
https://www.matrixscientific.com/061269.html
MSDS
https://www.matrixscientific.com/061269.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service