Skip to Content
Merck
All Photos(1)

Key Documents

KEY023878472

Key Organics / BIONET

2-benzoyl-3-(bromomethyl)-1-benzofuran

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
315.17
MDL number:

mp

104 - 106

Iupac Name

[3-(bromomethyl)-1-benzofuran-2-yl](phenyl)methanone

Inchi Code

1S/C16H11BrO2/c17-10-13-12-8-4-5-9-14(12)19-16(13)15(18)11-6-2-1-3-7-11/h1-9H,10H2

InChI key

IZKOJVVZRLUAKT-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/3J-584S

Melting Point

104 - 106

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/3J-584S


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service