Skip to Content
Merck
All Photos(1)

Key Documents

ENAH93E754BD

Enamine

dicyclohexyl[2-(phenanthren-9-yl)phenyl]phosphane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
450.6
MDL number:

mp

141-142

Iupac Name

dicyclohexyl(2-(phenanthren-9-yl)phenyl)phosphane

Inchi Code

1S/C32H35P/c1-3-14-25(15-4-1)33(26-16-5-2-6-17-26)32-22-12-11-21-30(32)31-23-24-13-7-8-18-27(24)28-19-9-10-20-29(28)31/h7-13,18-23,25-26H,1-6,14-17H2

InChI key

XVHJMCHKRAOXKF-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-22195496

Melting Point

141-142

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-22195496


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service