Skip to Content
Merck
All Photos(1)

Key Documents

ENAH0424C9C6

Enamine

N-methyl-N-(prop-2-en-1-yl)carbamoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
133.58
MDL number:

Iupac Name

allyl(methyl)carbamic chloride

Inchi Code

1S/C5H8ClNO/c1-3-4-7(2)5(6)8/h3H,1,4H2,2H3

InChI key

UEKTZPNFEQJPLZ-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-1612229

MSDS

https://www.enaminestore.com/catalog/EN300-1612229


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service