Skip to Content
Merck
All Photos(1)

Documents

CIAH987ED9D7

ChemScene LLC (Preferred Partner)

2,6-Dibromo-4,8-bis(n-octyloxy)benzo[1,2-b:4,5-b']-dithiophene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
604.51
MDL number:

bp

632.3+-50.0 C at 760 mmHg

mp

61.0-65.0 C

Iupac Name

2,6-dibromo-4,8-bis(octyloxy)benzo[1,2-b:4,5-b']dithiophene

Inchi Code

1S/C26H36Br2O2S2/c1-3-5-7-9-11-13-15-29-23-19-17-21(27)32-26(19)24(20-18-22(28)31-25(20)23)30-16-14-12-10-8-6-4-2/h17-18H,3-16H2,1-2H3

InChI key

GXGDTYKIKNMZTM-UHFFFAOYSA-N

storage temp.

4C

Purity

97%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/1294515-75-5.html

Boiling point

632.3+-50.0 C at 760 mmHg

Melting Point

61.0-65.0 C

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0167971.pdf


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service