Skip to Content
Merck
All Photos(1)

Key Documents

C12H316CEDB3

Chemcia Scientific, LLC

2-Chloro-5-fluoro-4-nitro-benzoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
233.58
MDL number:

Iupac Name

methyl 2-chloro-5-fluoro-4-nitrobenzoate

Inchi Code

1S/C8H5ClFNO4/c1-15-8(12)4-2-6(10)7(11(13)14)3-5(4)9/h2-3H,1H3

InChI key

GTHCSQMSHAUBMM-UHFFFAOYSA-N

storage temp.

0-5

Purity

97%

Country of Origin

US

Shipping Temperature

Ambient

Physical Form

Light Yellow Solid

Product Link

http://www.chemcia.com/product_fl02.asp?id=4640

MSDS

http://www.chemcia.com/product_fl02.asp?id=4640


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service