ATE371592366
AstaTech, Inc.
2,6-DIFLUORO-4-METHOXYPHENYLACETIC ACID
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
138-141 C
Iupac Name
(2,6-difluoro-4-methoxyphenyl)acetic acid
Inchi Code
1S/C9H8F2O3/c1-14-5-2-7(10)6(4-9(12)13)8(11)3-5/h2-3H,4H2,1H3,(H,12,13)
InChI key
KXWRAINJKJTUDG-UHFFFAOYSA-N
storage temp.
2-8 C
Purity
95%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
White to Yellow Solid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=12406
Melting Point
138-141 C
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=CL8751
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service