Skip to Content
Merck
All Photos(1)

Key Documents

APO497443788

Apollo Scientific Ltd

1,2-Dimethyl-1,4,5,6-tetrahydropyrimidine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
£16.50

Vendor SKU: OR13592-25G

£16.50


Ships in 5 days - from Aldrich Partner.


Select a Size

Change View
25 G
£16.50

About This Item

CAS Number:
Molecular Weight:
112.17
MDL number:

Vendor SKU: OR13592-25G

£16.50


Ships in 5 days - from Aldrich Partner.

mp

73

density

0.98

Iupac Name

1,2-dimethyl-1,4,5,6-tetrahydropyrimidine

Inchi Code

1S/C6H12N2/c1-6-7-4-3-5-8(6)2/h7H,1,3-5H2,2H3

InChI key

AVYDWKSWNZIAIW-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/12-dimethyl-1456-tetrahydropyrimidine

Flash Point

125

Melting Point

73

density

0.98

Pictograms

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/12-dimethyl-1456-tetrahydropyrimidine


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service