Skip to Content
Merck
All Photos(1)

Key Documents

APO456793379

Apollo Scientific Ltd

3-Bromo-6,8-dichloroimidazo[1,2-a]pyridine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
£18.70

Vendor SKU: OR3675-100MG

£18.70


Ships in 5 days - from Aldrich Partner.


Select a Size

Change View
100 MG
£18.70

About This Item

CAS Number:
Molecular Weight:
265.92
MDL number:

Vendor SKU: OR3675-100MG

£18.70


Ships in 5 days - from Aldrich Partner.

bp

111-113

Iupac Name

3-bromo-6,8-dichloroimidazo[1,2-a]pyridine

Inchi Code

1S/C7H3BrCl2N2/c8-6-2-11-7-5(10)1-4(9)3-12(6)7/h1-3H

InChI key

CJLKTLUNUJRVNA-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid-Powder

Product Link

https://store.apolloscientific.co.uk/product/3-bromo-68-dichloroimidazo12-apyridine

Boiling point

111-113

Pictograms

Exclamation mark

Signal Word

Warning

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/3-bromo-68-dichloroimidazo12-apyridine


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service