Skip to Content
Merck
All Photos(1)

Key Documents

APO455838318

Apollo Scientific Ltd

2,6-Difluoronicotinaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
143.09
MDL number:

bp

19-20

density

1.378

Iupac Name

2,6-difluoronicotinaldehyde

Inchi Code

1S/C6H3F2NO/c7-5-2-1-4(3-10)6(8)9-5/h1-3H

InChI key

ZYCCUPSLEFLBNR-UHFFFAOYSA-N

storage temp.

Store at -20C

Purity

97%

Country of Origin

GB

Shipping Temperature

Shipped with Ice Packs

Physical Form

Liquid-Clear

Product Link

https://store.apolloscientific.co.uk/product/26-difluoronicotinaldehyde

Synonym

2,6-Difluoropyridine-3-carboxaldehyde, 2,6-Difluoro-3-formylpyridine

Flash Point

64-66/15mm

Boiling point

19-20

density

1.378

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/26-difluoronicotinaldehyde


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service