APO455835359
Apollo Scientific Ltd
2-(Difluoromethoxy)bromobenzene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
>110
density
1.6
Iupac Name
2-bromophenyl difluoromethyl ether
Inchi Code
1S/C7H5BrF2O/c8-5-3-1-2-4-6(5)11-7(9)10/h1-4,7H
InChI key
VHEICCMNYWNFQX-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Purity
97%
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Liquid
Product Link
https://store.apolloscientific.co.uk/product/2-difluoromethoxybromobenzene
Synonym
2-Bromo-alpha,alpha-difluoroanisole, 2-Bromophenyl difluoromethyl ether, 1-Bromo-2-(difluoromethoxy)benzene
Flash Point
85-87/15mm
Melting Point
>110
density
1.6
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/2-difluoromethoxybromobenzene
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service