APO455834039
Apollo Scientific Ltd
1-Fluoro-2-iodoethane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
none
density
2.136
Iupac Name
1-fluoro-2-iodoethane
Inchi Code
1S/C2H4FI/c3-1-2-4/h1-2H2
InChI key
LVYJIIRJQDEGBR-UHFFFAOYSA-N
storage temp.
Store at 2-8C
Purity
98%
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Liquid
Product Link
https://store.apolloscientific.co.uk/product/1-fluoro-2-iodoethane
Synonym
2-Fluoro-1-iodoethane, 2-Iodoethyl fluoride, 2-Fluoroethyl iodide
Flash Point
98-102
Melting Point
none
density
2.136
Signal Word
Danger
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/1-fluoro-2-iodoethane
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service