APO455829241
Apollo Scientific Ltd
5-Carboxythiophene-2-boronic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
bp
135-136
Iupac Name
5-(dihydroxyboryl)-2-thiophenecarboxylic acid
Inchi Code
1S/C5H5BO4S/c7-5(8)3-1-2-4(11-3)6(9)10/h1-2,9-10H,(H,7,8)
InChI key
OQGIKNPOYTVNNF-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Solid
Product Link
https://store.apolloscientific.co.uk/product/5-carboxythiophene-2-boronic-acid
Boiling point
135-136
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/5-carboxythiophene-2-boronic-acid
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service