Skip to Content
Merck
All Photos(1)

Key Documents

APO455827029

Apollo Scientific Ltd

4-(Piperidin-4-yl)pyridine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
£39.60

Vendor SKU: OR13006-250MG

£39.60


Ships in 5 days - from Aldrich Partner.


Select a Size

Change View
250 MG
£39.60

About This Item

CAS Number:
Molecular Weight:
162.23
MDL number:

Vendor SKU: OR13006-250MG

£39.60


Ships in 5 days - from Aldrich Partner.

bp

86-88

Iupac Name

4-(4-pyridinyl)piperidine

Inchi Code

1S/C10H14N2/c1-5-11-6-2-9(1)10-3-7-12-8-4-10/h1-2,5-6,10,12H,3-4,7-8H2

InChI key

RGBWBVGQZFJTEO-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/4-piperidin-4-ylpyridine

Boiling point

86-88

Pictograms

Exclamation mark

Signal Word

Warning

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/4-piperidin-4-ylpyridine


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service