Skip to Content
Merck
All Photos(1)

Key Documents

APO455825573

Apollo Scientific Ltd

1-(Naphth-1-yl)piperazine dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
£64.90
1 G
£162.80

Vendor SKU: OR0079-250MG

£64.90


Ships in 5 days - from Aldrich Partner.


Select a Size

Change View
250 MG
£64.90
1 G
£162.80

About This Item

CAS Number:
Molecular Weight:
285.22
MDL number:

Vendor SKU: OR0079-250MG

£64.90


Ships in 5 days - from Aldrich Partner.

bp

>260

Iupac Name

1-(1-naphthyl)piperazine dihydrochloride

Inchi Code

1S/C14H16N2.2ClH/c1-2-6-13-12(4-1)5-3-7-14(13)16-10-8-15-9-11-16;;/h1-7,15H,8-11H2;2*1H

InChI key

ZGMNVJGEEHEXIQ-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/1-naphth-1-ylpiperazine-dihydrochloride

Boiling point

>260

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/1-naphth-1-ylpiperazine-dihydrochloride


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service