Iupac Name
1-bromo-2-(ethylsulfanyl)benzene
Inchi Code
1S/C8H9BrS/c1-2-10-8-6-4-3-5-7(8)9/h3-6H,2H2,1H3
InChI key
REUXZHQDXRDRII-UHFFFAOYSA-N
Purity
Country of Origin
Physical Form
Product Link
MSDS
https://aobchem.com/pub/media/files/sds/27765.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service