Iupac Name
2-fluoro-6-iodophenol
Inchi Code
1S/C6H4FIO/c7-4-2-1-3-5(8)6(4)9/h1-3,9H
InChI key
ZFARONFWYBCKDO-UHFFFAOYSA-N
storage temp.
Keep in dark place,Sealed in dry,2-8C
Purity
97%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
Yellow to brown liquid or Solid
Product Link
https://www.ambeed.com/products/28177-50-6.html
Signal Word
Warning
Hazard Statements
Precautionary statements
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service