771333
Diethyl 4-(trifluoromethyl)benzylphosphonate
97%
About This Item
Assay
97%
form
liquid
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
refractive index
n20/D 1.456
density
1.227 g/mL at 25 °C
SMILES string
O=P(OCC)(OCC)CC1=CC=C(C(F)(F)F)C=C1
InChI
1S/C12H16F3O3P/c1-3-17-19(16,18-4-2)9-10-5-7-11(8-6-10)12(13,14)15/h5-8H,3-4,9H2,1-2H3
InChI key
NTQKMIQYHZLRSW-UHFFFAOYSA-N
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service