Assay
98%
mp
84-87 °C (lit.)
SMILES string
Oc1c(Cl)cc(Cl)c(Cl)c1Br
InChI
1S/C6H2BrCl3O/c7-4-5(10)2(8)1-3(9)6(4)11/h1,11H
InChI key
IFXJBIXHNZHYGK-UHFFFAOYSA-N
General description
6-bromo-2,4,5-trichlorophenol (BrTriClP), also known as 2-bromo-3,4,6-trichlorophenol, is an brominated persistent organic pollutant. Its molecular structure has been analyzed based on its X-ray absorption fine structure (EXAFS) spectroscopy spectrum.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
?-Iodination of enaminones with bis(pyridine)iodonium(I) tetrafluoroborate
Tetrahedron Letters, Volume 38, Issue 48, 8397-8400 (1997)
Journal of medicinal chemistry, 48(6), 1709-1712 (2005-03-18)
A series of novel tetracyclic tetrahydrofuran derivatives was prepared and evaluated for its potential psychotropic properties. In vitro affinities for multiple dopaminergic, serotonergic, alpha-adrenergic, and histamine receptors and for the norepinephrine transporter as well as the ED(50) values obtained in
Chemistry (Weinheim an der Bergstrasse, Germany), 10(17), 4206-4213 (2004-09-08)
The use of bis(pyridine)iodonium tetrafluoroborate (IPy(2)BF(4)) as an oxidizing agent towards different types of alcohols is reported. The observed reactivity involves different reaction pathways, as a function both of the structures of the starting materials and of the experimental conditions.
Analytical and bioanalytical chemistry, 390(3), 921-928 (2007-11-14)
X-ray absorption fine structure (EXAFS) spectroscopy spectra were collected for three brominated persistent pollutants: 6-bromo-2,4,5-trichlorophenol (BrTriClP), pentabromophenol (PentaBrP) and 3,3',5,5'-tetrabromobisphenol A (TBBA). The substances were selected to be symmetrical (BrTriClP and TBBA) or asymmetrical (PentaBrP) with respect to the atomic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service