Skip to Content
Merck
All Photos(1)

Documents

530018

Sigma-Aldrich

3-Bromo-2-methylaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH3)C6H3NH2
CAS Number:
Molecular Weight:
186.05
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.619 (lit.)

bp

248 °C (lit.)

density

1.51 g/mL at 25 °C (lit.)

SMILES string

Cc1c(N)cccc1Br

InChI

1S/C7H8BrN/c1-5-6(8)3-2-4-7(5)9/h2-4H,9H2,1H3

InChI key

IILVSKMKMOJHMA-UHFFFAOYSA-N

General description

3-Bromo-2-methylaniline is a 3-halo-2-methylaniline derivative.

Application

3-Bromo-2-methylaniline may be used in the preparation of :
  • 3,4-dibromo-2-methylaniline
  • racemic 3,9-dibromo-4,10-dimethyl-6H,12H,5,11-methanodibenzodiazocine, a racemic derivative of Tröger′s base
  • 4-bromo-1H-indazole

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, Resolution, and Absolute Configuration of Difunctionalized Troger's Base Derivatives.
Kiehne U, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 14(14), 4246-4255 (2008)
A practical synthesis of a PI3K inhibitor under noncryogenic conditions via functionalization of a lithium triarylmagnesiate intermediate
Tian Q, et al.
Organic Process Research & Development, 17(1), 97-107 (2013)
Efficient and Complementary Methods Offering Access to Synthetically Valuable 1, 2-Dibromobenzenes.
Diemer V, et al.
European Journal of Organic Chemistry, 2011(2), 327-340 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service