419850
1,2-Bis(dimethylsilyl)benzene
98%
Synonym(s):
o-Phenylenebis(dimethylsilane)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H4[SiH(CH3)2]2
CAS Number:
Molecular Weight:
194.42
Beilstein:
2937027
MDL number:
UNSPSC Code:
12352000
Assay
98%
refractive index
n20/D 1.51 (lit.)
bp
101 °C/13 mmHg (lit.)
density
0.898 g/mL at 25 °C (lit.)
SMILES string
C[SiH](C)c1ccccc1[SiH](C)C
InChI
1S/C10H18Si2/c1-11(2)9-7-5-6-8-10(9)12(3)4/h5-8,11-12H,1-4H3
InChI key
QAUCEYVYCBYVDK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
Packaged in glass bottles
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Organometallics, 11, 2639-2639 (1992)
Bulletin de la Societe Chimique De France, 129, 667-667 (1993)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service