Skip to Content
Merck
All Photos(1)

Documents

418153

Sigma-Aldrich

2,9,16,23-Tetraphenoxy-29H,31H-phthalocyanine

Dye content 98 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H34N8O4
CAS Number:
Molecular Weight:
882.92
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

composition

Dye content, 98%

mp

195 °C (dec.) (lit.)

λmax

668 nm
703 nm (2nd)

SMILES string

O(c1ccccc1)c2ccc3c4nc(nc5[nH]c(nc6nc(nc7[nH]c(n4)c8cc(Oc9ccccc9)ccc78)c%10cc(Oc%11ccccc%11)ccc6%10)c%12cc(Oc%13ccccc%13)ccc5%12)c3c2

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service