Skip to Content
Merck
All Photos(1)

Documents

407526

Sigma-Aldrich

Pentaerythritol diacrylate monostearate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16CO2CH2C(CH2O2CCH=CH2)2CH2OH
CAS Number:
Molecular Weight:
510.70
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

solid

contains

500 ppm monomethyl ether hydroquinone as inhibitor

mp

29-31 °C (lit.)

density

1.018 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OCC(CO)(COC(=O)C=C)COC(=O)C=C

InChI

1S/C29H50O7/c1-4-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-28(33)36-25-29(22-30,23-34-26(31)5-2)24-35-27(32)6-3/h5-6,30H,2-4,7-25H2,1H3

InChI key

UXRNWUYCCQFHIH-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Schwabe et al.
International journal of pharmaceutics, 526(1-2), 178-187 (2017-05-01)
RNA interference (RNAi) is a promising technique to treat severe diseases on a pre-protein level. We and others postulate that the release of nanoparticle-complexed small interfering RNA (siRNA) from implanted biomaterials could provide structural support for tissue repair, combined with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service