286567
Mephenesin
98%
Synonym(s):
3-(2-Methylphenoxy)-1,2-propanediol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3C6H4OCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
182.22
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Assay
98%
form
powder
bp
153-154 °C/4 mmHg (lit.)
mp
69-71 °C (lit.)
SMILES string
Cc1ccccc1OCC(O)CO
InChI
1S/C10H14O3/c1-8-4-2-3-5-10(8)13-7-9(12)6-11/h2-5,9,11-12H,6-7H2,1H3
InChI key
JWDYCNIAQWPBHD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
S Bertrand et al.
The European journal of neuroscience, 11(2), 583-592 (1999-03-03)
The in vitro newborn rat isolated brain stem/spinal cord preparation was used to study the involvement of presynaptic inhibition in the control of the synaptic locomotor drive. The recording chamber was partitioned with Vaseline walls to separate the L1-L2 locomotor
C Beltran et al.
Annales de dermatologie et de venereologie, 136(10), 709-712 (2009-10-06)
In most cases, acute generalized exanthematous pustulosis (AGEP) is a drug reaction. Even if several systemic drugs have been reported to be a causative agent, this clinical case has rarely been observed with a topical treatment. We report here a
M A Shuker et al.
British journal of pharmacology, 111(3), 951-955 (1994-03-01)
1. Previous studies have suggested that a series of drugs modelled on part of the strychnine molecule interfere with the development of high pressure neurological syndrome (HPNS) and it was presumed that this effect was via an action on inhibitory
B Nickel et al.
Arzneimittel-Forschung, 40(8), 909-911 (1990-08-01)
The influence of the skeletal muscle tone by flupirtine (D-9998, Katadolon; CAS 56995-20-1), some selected analgesics and muscle relaxants was investigated in conscious rats after intraperitoneal administration. Benzodiazepines (diazepam and tetrazepam), baclofen, dantrolene and mephenesine reduced the tone of the
Shohei Yamamoto et al.
Brain research, 1082(1), 115-123 (2006-03-02)
Nociceptive signals are transmitted to the spinal dorsal horn via primary afferent fibers, and the signals induce withdrawal reflexes by activating spinal motoneurons in the ventral horn. Therefore, nociceptive stimuli increase motoneuronal firing and ventral root discharges. This study was
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service