Assay
99.9%
form
granular
density
8.8 g/mL at 25 °C (lit.)
SMILES string
[Ho]
InChI
1S/Ho
InChI key
KJZYNXUDTRRSPN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service