ADVH49B82623
AChemBlock
methyl 4-[(E)-hydroxyiminomethyl]benzoate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl 4-[(E)-(hydroxyimino)methyl]benzoate
Inchi Code
1S/C9H9NO3/c1-13-9(11)8-4-2-7(3-5-8)6-10-12/h2-5H,6H2,1H3
InChI key
LCTOGOXOYKDRNN-UHFFFAOYSA-N
storage temp.
Store at 0-8 C
Purity
97%
Country of Origin
CN
Physical Form
white solid
Product Link
https://www.achemblock.com/s78731-methyl-4-e-hydroxyiminomethyl-benzoate.html
Synonym
methyl (e)-4-((hydroxyimino)methyl)benzoate
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.achemblock.com/s78731-methyl-4-e-hydroxyiminomethyl-benzoate.html
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service