Skip to Content
Merck
All Photos(1)

Key Documents

AABH97D32053

AA BLOCKS, INC.

(3S)-N-isopropyl-N-methyl-pyrrolidin-3-amine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
142.24
MDL number:

Iupac Name

(3S)-N-isopropyl-N-methyl-3-pyrrolidinamine

Inchi Code

1S/C8H18N2/c1-7(2)10(3)8-4-5-9-6-8/h7-9H,4-6H2,1-3H3/t8-/m0/s1

InChI key

NSMOKJIWGMOVNM-QMMMGPOBSA-N

storage temp.

2~8 C

Purity

97%

Country of Origin

CN

Product Link

https://www.aablocks.com/prod/1018443-33-8

Synonym

(S)-N-Isopropyl-N-methylpyrrolidin-3-amine 2HCl

MSDS

https://www.aablocks.com/downloadSds?cas=1018443-33-8


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service