Skip to Content
Merck
All Photos(1)

Key Documents

5.04841

Sigma-Aldrich

5,7-Dichlorokynurenic Acid, Sodium Salt

Synonym(s):

5,7-Dichlorokynurenic Acid, Sodium Salt, 5,7-Dichloro-4-hydroxyquinoline-2-c-arboxylic acid sodium salt, DCKA

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
£79.70

£79.70


Estimated to ship on28 May 2025Details


Request a Bulk Order

Select a Size

Change View
10 MG
£79.70

About This Item

Empirical Formula (Hill Notation):
C10H4Cl2NNaO3
CAS Number:
Molecular Weight:
280.04
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

£79.70


Estimated to ship on28 May 2025Details


Request a Bulk Order

Assay

≥99% (HPLC)

Quality Level

form

solid

potency

79 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 10 mM
1 M NaOH: 100 mM

storage temp.

2-8°C

SMILES string

[Na+].Clc1c2c(nc(cc2O)C(=O)[O-])cc(c1)Cl

InChI

1S/C10H5Cl2NO3.Na/c11-4-1-5(12)9-6(2-4)13-7(10(15)16)3-8(9)14;/h1-3H,(H,13,14)(H,15,16);/q;+1/p-1

InChI key

VPRPMJHKWHCUFW-UHFFFAOYSA-M

General description

A very potent, water-soluble antagonist of the NMDA receptors complex acting at the strychnine-insensitive glycine binding site (Ki = 79 nM vs. [3H]-glycine).

Biochem/physiol Actions

Primary Target
NMDA receptors

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Baron, B., et al. 1991. Eur. J. Pharmacol.206, 149.
Baron, B., et al. 1990. Mol. Pharmacol.38, 554.
Mugnaini, M., et al. 1998. J. Recept. Signal Transduct. Res.18, 91.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service