Merck
HomeWestern BlottingAlkaline Phosphatase
Alkaline Phosphatase
Alkaline Phosphatase — Assays, Molecular Weight, Substrates, and Structure
We offer a broad range of alkaline phosphatase (ALP/ALKP) preparations optimized for conjugation to antibodies and other proteins for ELISA, Western blotting, and histochemical detection. Our BioUltra Grade Alkaline Phosphatase has a very high specific activity making it particularly useful for protein labeling when high sensitivity is required.
Western Blotting
Western Blotting and Immunohistochemistry
Figure 1.FLAG-BAP control protein detected with anti-FLAG antibody and ALP-labeled secondary antibody and stained with BCIP/NBT substrate (using ProteoQuest Colorimetric Western Blotting Kit, B6404).
Immunohistochemistry
Colorimetric immunohistochemical detection of Alkaline Phosphatase
Figure 2.Formalin-fixed, paraffin-embedded section of human tonsil stained with Monoclonal Anti-Human IgA and an Anti-Mouse IgG ALP Conjugate using SIGMAFAST™ Fast Red TR/Naphthol AS-MX Tablets (Cat. No. F4523) as substrate. 40x
Our products are also used to dephosphorylate casein and other proteins. ALKP may be used to dephosphorylate the 5'-termini of DNA or RNA to prevent self-ligation. DNA or RNA can also be tagged with radiolabeled phosphate (via T4 polynucleotide kinase) after dephosphorylation with ALKP.
Bovine Alkaline Phosphatase
Bovine intestinal ALP is a dimeric, membrane-derived glycoprotein.1-3 At least three isoforms exist, which typically possess two N-linked and one or more O-linked glycans per monomer.2 The enzyme requires zinc, and magnesium or calcium divalent ions for activity.4
The enzyme has a broad specificity for phosphate esters of alcohols, amines, pyrophosphate, and phenols. It is routinely used to dephosphorylate proteins and nucleic acids.5-7
KM: 1.5 × 10–3 M (p-Nitrophenyl phosphate), 19 × 10–3 M (phosphoenolpyruvate)
Molecular weight:2,3 140–160 kDa (dimer MW)
E2781%: 7.6–10.5
Isoelectric point:4,8,9 Isozymes with a pI range of 4.4–5.8
pH Optimum: The enzyme is most stable in the pH range 7.5–9.5.3 The pH optimum for enzymatic activity is pH 8-10. The pH optimum will change depending upon substrate, substrate concentration, and ionic concentration.8 The enzyme activity for this product is determined at pH 9.8 (diethanolamine buffer enzyme assay).
Graphs showing pH Optimum of Alkaline Phosphatase
Temperature Effects on Activity and Stability
Temperature Effects on the Activity and Stability of Alkaline Phosphatase
Inhibitors of Bovine Alkaline Phosphatase
Chelating agents, arsenate, cysteine, iodine, inorganic phosphate, pyrophosphate, diisopropyl phosphate, triphenylphosphate, diisopropyl fluorophosphate, and L-phenylalanine.9,10
Levamisole (Catalog Number L9756) is typically used to inhibit endogenous ALKP activity, while only slightly inhibiting the intestinal enzyme.11,12
Assay Protocols
Unit Definition: One DEA unit will hydrolyze 1 µmole of 4-nitrophenyl phosphate per minute at pH 9.8 at 37 °C. Approximately 3 DEA units equal one Glycine unit.
Alkaline Phosphatase Reaction with Substrate pNPP
ALP is also used to determine the effectiveness of pasteurization on dairy products such as milk and cheese. ALP denatures when subjected to temperatures of approximately 72 °C for at least 15 seconds. Less extreme temperature conditions have been shown to destroy most milk-born pathogens. Therefore, ALP activity can be used to measure the effectiveness of pasteurization. In the past, two methods have been used to check for ALP activity after pasteurization, the first being the Scharer Test, which liberates and measures phenol. The second method is known as the Aschaffenberg & Mullen Test. This method measures the release of p-nitrophenol. With both tests negative results signify no ALP activity and the destruction of most milk-born pathogens. Recently new more sensitive techniques have been developed based on the same ALP activity principle.
References
1.
Hsu HH, Munoz PA, Barr J, Oppliger I, Morris DC, Vaananen HK, Tarkenton N, Anderson HC. 1985. Purification and partial characterization of alkaline phosphatase of matrix vesicles from fetal bovine epiphyseal cartilage. Purification by monoclonal antibody affinity chromatography. J. Biol. Chem..(260):1826-31.
2.
NEUMANN H, LUSTIG A. 1980. The Activation of Alkaline Phosphatase by Effector Molecules. A Combined Kinetic and Hydrodynamic Study. Eur J Biochem. 109(2):475-480. https://doi.org/10.1111/j.1432-1033.1980.tb04818.x
3.
Fosset M, Chappelet-Tordo D, Lazdunski M. 1974. Intestinal alkaline phosphatase. Physical properties and quaternary structure. Biochemistry. 13(9):1783-1788. https://doi.org/10.1021/bi00706a001
4.
Besman M, Coleman JE. 1985. Isozymes of bovine intestinal alkaline phosphatase. J Biol. Chem..(260):11190-3.
5.
Fernley H. 1971. 18 Mammalian Alkaline Phosphatases.417-447. https://doi.org/10.1016/s1874-6047(08)60378-9
6.
Morton RK. 1955. The substrate specificity and inhibition of alkaline phosphatases of cow's milk and calf intestinal mucosa. 61(2):232-240. https://doi.org/10.1042/bj0610232
7.
Morton RK. 1955. The action of purified alkaline phosphatases on di- and tri-phosphopyridine nucleotides. 61(2):240-244. https://doi.org/10.1042/bj0610240
8.
Latner AL, Parsons M, Skillen AW. 1970. Isoelectric focusing of alkaline phosphatases from human kidney and calf intestine. Enzymologia. 40(1):1-7.
9.
Lazdunski M, Brouillard J, Ouellet L. 1965. ÉTUDE DES EFFETS ÉLECTROSTATIQUES SUR LE MÉCANISME D'ACTION CATALYTIQUE DE LA PHOSPHATASE ALCALINE INTESTINALE DE VEAU. Can. J. Chem.. 43(8):2222-2235. https://doi.org/10.1139/v65-300
10.
Harlow E, Lane D. 1988. Antibodies: A Laboratory Manual. 1. New York: Cold Spring Harbor Larboratory Press.
11.
STAGNI N, VITTUR F, DEBERNARD B. 1983. Solubility properties of alkaline phosphatase from matrix vesicles. Biochimica et Biophysica Acta (BBA) - General Subjects. 761(3):246-251. https://doi.org/10.1016/0304-4165(83)90072-7
12.
van Belle H. 1972. Kinetics and inhibition of alkaline phosphatases from canine tissues. Biochimica et Biophysica Acta (BBA) - Enzymology. 289(1):158-168. https://doi.org/10.1016/0005-2744(72)90118-0
13.
Roger DM. 1938. Practical Experience with the Scharer Rapid Field Test for Pasteurization. Am J Public Health Nations Health. 28(11):1325-1327. https://doi.org/10.2105/ajph.28.11.1325
E. coli Alkaline Phosphatase
E. coli ALKP is a dimeric, non-glycosylated protein assumed to reside mainly in the periplasmic space of E. coli. At least three isoforms exist. The enzyme requires zinc and is further activated by magnesium ions for activity.1-3 Like the bovine enzyme, the E. coli enzyme has a broad specificity for phosphate esters.1
KM: 0.02 × 10–3 M (p-Nitrophenyl phosphate)4
Molecular weight:2 89 kDa (Dimer MW)
E2781%: 7.6–10.52
Isoelectric point:5 Isozymes with a pI range of 4.5
pH Optimum: 8.05
1.
Reid TW, Wilson IB. 1971. 17 E. coli Alkaline Phosphatase.373-415. https://doi.org/10.1016/s1874-6047(08)60377-7
2.
Anderson RA, Vallee BL. 1975. Cobalt(III), a probe of metal binding sites of Escherichia coli alkaline phosphatase.. Proceedings of the National Academy of Sciences. 72(1):394-397. https://doi.org/10.1073/pnas.72.1.394
3.
Savchenko A, Wang W, Vieille C, Zeikus J. 2001. [26] Alkaline phosphatase from Thennotoga neapolitana.298-305. https://doi.org/10.1016/s0076-6879(01)31067-4
4.
Boulanger RR, Kantrowitz ER. 2003. Characterization of a MonomericEscherichia coliAlkaline Phosphatase Formed upon a Single Amino Acid Substitution. J. Biol. Chem.. 278(26):23497-23501. https://doi.org/10.1074/jbc.m301105200
5.
Garen A, Levinthal C. 1960. A fine-structure genetic and chemical study of the enzyme alkaline phosphatase of E. Coli I. Purification and characterization of alkaline phosphatase. Biochimica et Biophysica Acta. 38470-483. https://doi.org/10.1016/0006-3002(60)91282-8
Shrimp Alkaline Phosphatase
Shrimp ALKP is a dimeric protein with a molecular weight of approximately 58 kDa as determined by SDS-PAGE.
The Shrimp enzyme denatures at a lower temperature than other sources of the enzyme and is therefore recommended for dephosphorylation of DNA and RNA when a post-reaction heat inactivation step is required.
Shrimp Alkaline Phosphatase, Cat. No. A2237
Procedure for Dephosphorylation of DNA