Merck
  • Comprehensive functional characterization of cancer-testis antigens defines obligate participation in multiple hallmarks of cancer.
Comprehensive functional characterization of cancer-testis antigens defines obligate participation in multiple hallmarks of cancer.
Nature communications (2015-11-17)
Kimberly E Maxfield, Patrick J Taus, Kathleen Corcoran, Joshua Wooten, Jennifer Macion, Yunyun Zhou, Mark Borromeo, Rahul K Kollipara, Jingsheng Yan, Yang Xie, Xian-Jin Xie, Angelique W Whitehurst
ABSTRACT
Tumours frequently activate genes whose expression is otherwise biased to the testis, collectively known as cancer-testis antigens (CTAs). The extent to which CTA expression represents epiphenomena or confers tumorigenic traits is unknown. In this study, to address this, we implemented a multidimensional functional genomics approach that incorporates 7 different phenotypic assays in 11 distinct disease settings. We identify 26 CTAs that are essential for tumor cell viability and/or are pathological drivers of HIF, WNT or TGFβ signalling. In particular, we discover that Foetal and Adult Testis Expressed 1 (FATE1) is a key survival factor in multiple oncogenic backgrounds. FATE1 prevents the accumulation of the stress-sensing BH3-only protein, BCL-2-Interacting Killer (BIK), thereby permitting viability in the presence of toxic stimuli. Furthermore, ZNF165 promotes TGFβ signalling by directly suppressing the expression of negative feedback regulatory pathways. This action is essential for the survival of triple negative breast cancer cells in vitro and in vivo. Thus, CTAs make significant direct contributions to tumour biology.
Sigma-Aldrich
Anti-Smad2/3 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-FATE1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone 2-28-33, ascites fluid
Sigma-Aldrich
Anti-RNF183 antibody produced in rabbit, affinity isolated antibody