Merck
  • Suitability of urethane anesthesia for physiopharmacological investigations in various systems. Part 2: Cardiovascular system.
Suitability of urethane anesthesia for physiopharmacological investigations in various systems. Part 2: Cardiovascular system.
Experientia (1986-03-15)
C A Maggi, A Meli
ABSTRACT
Urethane produces a level of surgical anesthesia characterized by preservation of a number of cardiovascular reflexes. When the proper route of administration is used, and the use of unnecessarily high doses is avoided, urethane anesthesia appears to be suitable for a number of investigations at cardiovascular level. However in certain types of studies involving pharmacological stimulation of peripheral adrenoceptors urethane affects markedly the magnitude of the response under study.
Product Description
Supelco
Urethane, analytical standard
Sigma-Aldrich
Urethane, ≥99%
Sigma-Aldrich
Urethane, ≥99.0% (GC)