Merck
  • Mycobacterium avium subsp. avium and Mycobacterium neoaurum detection in an immunocompromised patient.
Mycobacterium avium subsp. avium and Mycobacterium neoaurum detection in an immunocompromised patient.
Epidemiology and infection (2013-07-12)
M Kaevska, J Sterba, J Svobodova, I Pavlik
ABSTRACT
Non-tuberculous mycobacteria are increasingly described as infectious agents in immunocompromised patients. A 17-year-old male patient suffering from secondary non-Hodgkin's lymphoma and treated with chemotherapeutic agents was admitted to hospital due to pleuropneumonia. Mycobacterium neoaurum was cultured repeatedly from his sputum and, Mycobacterium avium subsp. avium (M. a. avium) was detected by IS901 qPCR from detached fragments of his intestinal mucosa. We attempted to determine the possible sources of infection by analysing environmental samples from the closed oncology unit and conventional unit in the hospital, and from the patient's home residence and places which he frequented. The environment of the patient harboured mycobacteria (41 isolates in total); however, M. neoaurum was not recovered. M. a. avium was detected by qPCR in the environmental samples from a small flock of hens kept by his neighbour. Although it was not confirmed by DNA fingerprinting methods, the M. a. avium infection could have been acquired through the eating of incompletely cooked eggs.
Product Description
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Ciprofloxacin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Ciprofloxacin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Ciprofloxacin HCl, Pharmaceutical Secondary Standard; Certified Reference Material