Merck
  • Acute and long-term effects of nine chemicals on the Japanese medaka (Oryzias latipes).
Acute and long-term effects of nine chemicals on the Japanese medaka (Oryzias latipes).
Archives of environmental contamination and toxicology (1995-04-01)
G W Holcombe, D A Benoit, D E Hammermeister, E N Leonard, R D Johnson
ABSTRACT
Ninety-six-hour acute and 28-day larval survival and growth tests were conducted with nine organic chemicals, using the Japanese medaka (Oryzias latipes) as the test organism. The nine tested chemicals were allyl isothiocyanate, aniline, benzyl acetate, 4-chloroaniline, 2-chloroethanol, 2,4-diaminotoluene, 1,2-dibromoethane, 2,4-dichlorophenoxyacetic acid (2,4-D), and phenol. The derived 96-h LC50 values for medaka for all chemicals ranged from 0.077 mg/L for allyl isothiocyanate to 2,780 mg/L for 2,4-D. The chronic values for six of the nine chemicals tested ranged from 0.013 mg/L for allyl isothiocyanate to 42.5 mg/L for 2,4-D. Acute-to-chronic ratios for these six chemicals ranged from 1.4 for 2-chloroethanol to 70.9 for 2,4-D. Growth of medaka was significantly reduced in the lowest exposure concentration during 28-dy larval tests with aniline, 4-chloroaniline, and 2,4-diaminotoluene. The estimated maximum acceptable toxicant concentration was reported as less than the lowest exposure concentration of 4.6, 2.2 and 40.3 mg/L for tests with aniline, 4-chloroaniline and 2,4-diaminotoluene, respectively. Chronic values for 2-chloroethanol and medaka were 12.6 mg/L during an embryo-larval test and 22.1 mg/L during the 28-day larval test.
Product Description
Supelco
Benzyl acetate, analytical standard
Sigma-Aldrich
Benzyl acetate, ≥99%
Sigma-Aldrich
Benzyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl acetate, natural, ≥99%, FCC, FG