Merck
Microbial growth on 2-bromobutane.
Antonie van Leeuwenhoek (1980-01-01)
G T Sperl, J McKae
ABSTRACT
A member of the genus Arthrobacter was isolated which grew at the expense of 2-bromobutane as sole source of carbon and energy. Evidence is presented which suggests that the initial conversion of 2-bromobutane to 2-butanol is a spontaneous chemical hydrolysis and not mediated by the organism. Further evidence from oxygen consumption experiments indicates that 2-bromobutane is oxidized through 2-butanol, methyl ethyl ketone, ethyl acetate to acetate and ethanol. Results of experiments with cells grown on pathway intermediates reveal that the enzymes necessary for the oxidation of 2-butanol, methyl ethyl ketone, ethyl acetate, ethanol and acetaldehyde are not coordinately, but individually induced by their respective substrates.
Sigma-Aldrich
2-Bromobutane, 98%