Merck
  • Evidence for a critical micelle concentration of surfactants in hydrocarbon solvents.
Evidence for a critical micelle concentration of surfactants in hydrocarbon solvents.
Langmuir : the ACS journal of surfaces and colloids (2013-02-16)
Gregory N Smith, Paul Brown, Sarah E Rogers, Julian Eastoe
ABSTRACT
The concentration-dependent aggregation of two surfactants, anionic sodium dioctylsulfosuccinate (Aerosol OT or AOT) and nonionic pentaethylene glycol monododecyl ether (C12E5), has been studied in cyclohexane-D12 using small-angle neutron scattering (SANS). A clear monomer-to-aggregate transition has been observed for both surfactants, spherical inverse micelles for AOT and hank-like micelles for C12E5. This suggests that a critical micelle concentration exists for surfactants of these kinds in nonpolar solvents. The nature of the transition is different for the two surfactants. AOT aggregates are the same size and shape with decreasing concentration until a sharp critical micelle concentration, after which they cannot be detected. However, C12E5 aggregates gradually decrease in size. These differences demonstrate that the strength of the solvophobic effect can influence the formation of surfactant aggregates in nonaqueous solvents.
Sigma-Aldrich
Dioctyl sulfosuccinate sodium salt, ≥97%
Sigma-Aldrich
Docusate sodium salt, BioUltra, ≥99.0% (TLC)
Sigma-Aldrich
Docusate sodium salt, purum, ≥96.0% (TLC)
Sigma-Aldrich
Pentaethylene glycol monododecyl ether, BioXtra, ≥98.0% (GC)
Sigma-Aldrich
Docusate sodium salt, BioXtra, ≥99%
Sigma-Aldrich
Docusate sodium, meets USP testing specifications