Merck
  • In vivo efficacy studies of layer-by-layer nano-matrix bearing kaempferol for the conditions of osteoporosis: a study in ovariectomized rat model.
In vivo efficacy studies of layer-by-layer nano-matrix bearing kaempferol for the conditions of osteoporosis: a study in ovariectomized rat model.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2012-08-29)
Avinash Kumar, Girish K Gupta, Vikram Khedgikar, Jyoti Gautam, Priyanka Kushwaha, Bendangla Changkija, Geet K Nagar, Varsha Gupta, Ashwni Verma, Anil Kumar Dwivedi, Naibedya Chattopadhyay, Prabhat Ranjan Mishra, Ritu Trivedi
ABSTRACT
A prototype formulation based on layer-by-layer (LbL) nano-matrix was developed to increase bioavailability of kaempferol with improved retention in bone marrow to achieve enhanced bone formation. The layer-by-layer nano-matrix was prepared by sequential adsorption of biocompatible polyelectrolytes over the preformed kaempferol-loaded CaCO(3) template. The system was pharmaceutically characterized and evaluated for osteogenic activity in ovariectomized (OVx) rats. Data have been compared to the standard osteogenic agent parathyroid hormone (PTH). Single oral dose of kaempferol loaded LbL nano-matrix formulation increased bioavailability significantly compared to unformulated kaempferol. Three months of Formulated kaempferol administration to osteopenic rats increased plasma and bone marrow Kaempferol levels by 2.8- and 1.75-fold, respectively, compared to free Kaempferol. Formulated Kaempferol increased bone marrow osteoprogenitor cells, osteogenic genes in femur, bone formation rate, and improved trabecular micro-architecture. Withdrawal of Formulated kaempferol-in OVx rats resulted in the maintenance of bone micro-architecture up to 30days, whereas micro-architectural deterioration was readily observed in OVx rats treated with unformulated kaempferol-within 15days of withdrawal. The developed novel formulation has enhanced anabolic effect in osteopenic rats through increased stimulatory effect in osteoblasts. Treatment post-withdrawal sustenance of formulated kaempferol could become a strategy to enhance bioavailability of flavanoids.
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Supelco
Kaempferol, analytical standard
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)