Merck
  • Hexahomotrioxacalix[3]arene derivatives as ionophores for molecular recognition of dopamine, serotonin and phenylethylamine.
Hexahomotrioxacalix[3]arene derivatives as ionophores for molecular recognition of dopamine, serotonin and phenylethylamine.
Organic & biomolecular chemistry (2012-05-17)
Xin-Long Ni, Shofiur Rahman, Shi Wang, Cheng-Cheng Jin, Xi Zeng, David L Hughes, Carl Redshaw, Takehiko Yamato
ABSTRACT
The lower rim functionalized hexahomotrioxacalix[3]arene derivatives cone-3 and cone-5 bearing three benzyl and three N,N-diethyl-2-aminoethoxy groups, respectively, were synthesized from triol 1. Their complexation with 2-(3,4-dihydroxyphenyl)ethylamine (dopamine), 5-hydroxytryptamine (serotonin), and 2-phenylethylamine (phenethylamine), which have biologically important activities, has been studied by (1)H-NMR spectroscopy. The chemical shifts of the aromatic protons of the host and guest molecules and the up-field shifts of the ethyl protons of the guest molecules strongly suggest the formation of inclusion complexes in solution. The formation of the host-guest complexes is assisted by a hydrogen bond and/or an electrostatic interaction between the host and ammonium ion (RNH(3)(+)) of the guest. The structures of receptors cone-3 and cone-5 have been determined by X-ray crystallography.
Product Description
Supelco
2-Phenethylamine, analytical standard
Sigma-Aldrich
Phenethylamine, ≥99%
Sigma-Aldrich
Phenethylamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Phenethylamine, 99%
Sigma-Aldrich
2-Phenylethylamine hydrochloride, ≥98%